Learn More
Abnova™ Human EPPB9 Full-length ORF (AAH02944.1, 1 a.a. - 153 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00027077-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The B9 protein function is not known. Identification of a conserved mouse homolog and representation in the EST database indicates that this transcript does represent a gene. This gene is located within the Smith-Magenis syndrome region on chromosome 17. [provided by RefSeq]
Sequence: MATASPSVFLLMVNGQVESAQFPEYDDLYCKYCFVYGQDWAPTAGLEEGISQITSKSQDVRQALVWNFPIDVTFKSTNPYGWPQIVLSVYGPDVFGNDVVRGYGAVHVPFSPGRHKRTIPMFVPESTSKLQKFTSLCLVASSDLQAAPPTEDKEspecificaciones
AAH02944.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
43.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
B9D1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
27077 | |
EPPB9 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
B9/EPPB9 | |
B9D1 | |
Recombinant | |
wheat germ expression system |