Learn More
ACROBiosystems Human Ephrin-A4 / EFNA4 Protein, Fc Tag
Marca:  ACROBiosystems EA4-H5250-250UG
Detalles adicionales : Peso : 0.00001kg
 Ver más versiones de este producto
Código de producto. 15886597
  • 290.32€ / 250 microgramos
Fecha estimada de envío: 10-05-2024
para ver el stock
Descripción
Descripción
Ephrin-A4 (EFNA4) is also known as EPH-related receptor tyrosine kinase ligand 4 (LERK4), EPLG4, which belongs to the ephrin family. EFNA4 contains one ephrin RBD (ephrin receptor-binding) domain. EFNA4 is cell surface GPI-bound ligand for Eph receptors, a family of receptor tyrosine kinases which are crucial for migration, repulsion and adhesion during neuronal, vascular and epithelial development. EFNA4 binds promiscuously Eph receptors residing on adjacent cells, leading to contact-dependent bidirectional signaling into neighboring cells. EFNA4 may play a role in the interaction between activated B-lymphocytes and dendritic cells in tonsils.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 43.0 kDa. The protein migrates as 45-50 kDa and 66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Especificaciones
Especificaciones
AAI07484
250 μg
Leu 26 - Gly 171
EFNA4,EPLG4,LERK4,Ephrin-A4
Unconjugated
Fc Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
43.0 kDa
Human Ephrin-A4, Fc Tag (EA4-H5250) is expressed from human 293 cells (HEK293). It contains AA Leu 26 - Gly 171 (Accession # AAI07484).
-20°C
Ephrin-A4
Human
Tris with Glycine, Arginine and NaCl, pH7.5
HEK293
Bio-Markers and CD Antigens