missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human EPC1 Partial ORF (NP_079485, 695 a.a. - 798 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00080314-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: KGLHLSSTTPTALVHTSPSTAGSALLQPSNITQTSSSHSALSHQVTAANSATTQVLIGNNIRLTVPSSVATVNSIAPINARHIPRTLSAVPSSALKLAAAANCQEspecificaciones
NP_079485 | |
Liquid | |
80314 | |
EPC1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp781P2312/Epl1 | |
EPC1 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.18kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KGLHLSSTTPTALVHTSPSTAGSALLQPSNITQTSSSHSALSHQVTAANSATTQVLIGNNIRLTVPSSVATVNSIAPINARHIPRTLSAVPSSALKLAAAANCQ | |
RUO | |
EPC1 | |
Wheat Germ (in vitro) | |
GST |