missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ENTPD6 Full-length ORF (AAH25980.1, 1 a.a. - 483 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00000955-P01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16192861

  • 508.00€ / 25 microgramos
Fecha estimada de envío: 10-06-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

ENTPD6 is similar to E-type nucleotidases (NTPases). NTPases, such as CD39, mediate catabolism of extracellular nucleotides. ENTPD6 contains 4 apyrase-conserved regions which is characteristic of NTPases. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MKKGIRYETSRKTSYIFQPQHGPWQTRMRKISNHGSLRVAKVAYPLGLCVGVFIYVAYIKWHRATATQAFFSITRAAPGARWGQQAHSPLGTAADGHEVFYGIMFDAGSTGTRVHVFQFTRPPRETPTLTHETFKALKPGLSAYADDVEKSAQGIRELLDVAKQDIPFDFWKATPLVLKATAGLRLLPGEKAQKLLQKVKEVFKASPFLVGDDCVSIMNGTDEGVSAWITINFLTGSLKTPGGSSVGMLDLGGGSTQIAFLPRVEGTLQASPPGYLTALRMFNRTYKLYSYSYLGLGLMSARLAILGGVEGQPAKDGKELVSPCLSPSFKGEWEHAEVTYRVSGQKAAASLHELCAARVSEVLQNRVHRTEEVKHVDFYAFSYYYDLAAGVGLIDAEKGGSLVVGDFEIAAKYVCRTLETQPQSSPFSCMDLTYVSLLLQEFGFPRSKVLKLTRKIDNVETSWALGAIFHYIDSLNRQKSPAS
Especificaciones

Especificaciones

AAH25980.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
79.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CD39L2/DKFZp781G2277/DKFZp781K21102/FLJ36711/IL-6SAG/IL6ST2/NTPDase-6/dJ738P15.3
ENTPD6
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
955
ENTPD6 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MKKGIRYETSRKTSYIFQPQHGPWQTRMRKISNHGSLRVAKVAYPLGLCVGVFIYVAYIKWHRATATQAFFSITRAAPGARWGQQAHSPLGTAADGHEVFYGIMFDAGSTGTRVHVFQFTRPPRETPTLTHETFKALKPGLSAYADDVEKSAQGIRELLDVAKQDIPFDFWKATPLVLKATAGLRLLPGEKAQKLLQKVKEVFKASPFLVGDDCVSIMNGTDEGVSAWITINFLTGSLKTPGGSSVGMLDLGGGSTQIAFLPRVEGTLQASPPGYLTALRMFNRTYKLYSYSYLGLGLMSARLAILGGVEGQPAKDGKELVSPCLSPSFKGEWEHAEVTYRVSGQKAAASLHELCAARVSEVLQNRVHRTEEVKHVDFYAFSYYYDLAAGVGLIDAEKGGSLVVGDFEIAAKYVCRTLETQPQSSPFSCMDLTYVSLLLQEFGFPRSKVLKLTRKIDNVETSWALGAIFHYIDSLNRQKSPAS
RUO
ENTPD6
Wheat Germ (in vitro)
GST
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales