Learn More
Abnova™ Human ENTPD6 Full-length ORF (AAH25980.1, 1 a.a. - 483 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00000955-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Description
ENTPD6 is similar to E-type nucleotidases (NTPases). NTPases, such as CD39, mediate catabolism of extracellular nucleotides. ENTPD6 contains 4 apyrase-conserved regions which is characteristic of NTPases. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MKKGIRYETSRKTSYIFQPQHGPWQTRMRKISNHGSLRVAKVAYPLGLCVGVFIYVAYIKWHRATATQAFFSITRAAPGARWGQQAHSPLGTAADGHEVFYGIMFDAGSTGTRVHVFQFTRPPRETPTLTHETFKALKPGLSAYADDVEKSAQGIRELLDVAKQDIPFDFWKATPLVLKATAGLRLLPGEKAQKLLQKVKEVFKASPFLVGDDCVSIMNGTDEGVSAWITINFLTGSLKTPGGSSVGMLDLGGGSTQIAFLPRVEGTLQASPPGYLTALRMFNRTYKLYSYSYLGLGLMSARLAILGGVEGQPAKDGKELVSPCLSPSFKGEWEHAEVTYRVSGQKAAASLHELCAARVSEVLQNRVHRTEEVKHVDFYAFSYYYDLAAGVGLIDAEKGGSLVVGDFEIAAKYVCRTLETQPQSSPFSCMDLTYVSLLLQEFGFPRSKVLKLTRKIDNVETSWALGAIFHYIDSLNRQKSPASSpecifications
AAH25980.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
79.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CD39L2/DKFZp781G2277/DKFZp781K21102/FLJ36711/IL-6SAG/IL6ST2/NTPDase-6/dJ738P15.3 | |
ENTPD6 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
955 | |
ENTPD6 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MKKGIRYETSRKTSYIFQPQHGPWQTRMRKISNHGSLRVAKVAYPLGLCVGVFIYVAYIKWHRATATQAFFSITRAAPGARWGQQAHSPLGTAADGHEVFYGIMFDAGSTGTRVHVFQFTRPPRETPTLTHETFKALKPGLSAYADDVEKSAQGIRELLDVAKQDIPFDFWKATPLVLKATAGLRLLPGEKAQKLLQKVKEVFKASPFLVGDDCVSIMNGTDEGVSAWITINFLTGSLKTPGGSSVGMLDLGGGSTQIAFLPRVEGTLQASPPGYLTALRMFNRTYKLYSYSYLGLGLMSARLAILGGVEGQPAKDGKELVSPCLSPSFKGEWEHAEVTYRVSGQKAAASLHELCAARVSEVLQNRVHRTEEVKHVDFYAFSYYYDLAAGVGLIDAEKGGSLVVGDFEIAAKYVCRTLETQPQSSPFSCMDLTYVSLLLQEFGFPRSKVLKLTRKIDNVETSWALGAIFHYIDSLNRQKSPAS | |
RUO | |
ENTPD6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |