Learn More
Abnova™ Human ELMO2 Partial ORF (NP_573403.1, 89 a.a. - 188 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00063916-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene interacts with the dedicator of cyto-kinesis 1 protein. Similarity to a C. elegans protein suggests that this protein may function in phagocytosis of apoptotic cells and in cell migration. Alternative splicing results in multiple transcript variants encoding the same protein. [provided by RefSeq]
Sequence: MERTQSSNMETRLDAMKELAKLSADVTFATEFINMDGIIVLTRLVESGTKLLSHYSEMLAFTLTAFLELMDHGIVSWDMVSITFIKQIAGYVSQPMVDVSEspecificaciones
NP_573403.1 | |
Liquid | |
63916 | |
ELMO2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CED-12/CED12/ELMO-2/FLJ11656/KIAA1834 | |
ELMO2 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MERTQSSNMETRLDAMKELAKLSADVTFATEFINMDGIIVLTRLVESGTKLLSHYSEMLAFTLTAFLELMDHGIVSWDMVSITFIKQIAGYVSQPMVDVS | |
RUO | |
ELMO2 | |
Wheat Germ (in vitro) | |
GST |