Learn More
Abnova™ Human DSTYK Partial ORF (NP_056190, 80 a.a. - 189 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00025778-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a dual serine/threonine and tyrosine protein kinase which is expressed in multiple tissues. It is thought to function as a regulator of cell death. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: DVAETGLQAGQLSCISFPPKEEKYLQQIVDCLPCILILGQDCNVKCQLLNLLLGVQVLPTTKLGSEESCKLRRLRFTYGTQTRVSLALPGQYELVHTLVAHQGNWETIPEEspecificaciones
NP_056190 | |
Liquid | |
25778 | |
DSTYK (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DustyPK/HDCMD38P/KIAA0472/RIP5/RIPK5 | |
DSTYK | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DVAETGLQAGQLSCISFPPKEEKYLQQIVDCLPCILILGQDCNVKCQLLNLLLGVQVLPTTKLGSEESCKLRRLRFTYGTQTRVSLALPGQYELVHTLVAHQGNWETIPE | |
RUO | |
DSTYK | |
Wheat Germ (in vitro) | |
GST |