missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human DMRTA1 Partial ORF (NP_071443, 301 a.a. - 390 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00063951-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: ESGNESEWVKDLTSTKASLPTVSSRPRDPLDILTKIFPNYRRSRLEGILRFCKGDVVQAIEQVLNGKEHKPDNRNLANSEELENTAFQRAEspecificaciones
NP_071443 | |
Liquid | |
63951 | |
DMRTA1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DMO/MGC163307/MGC163309 | |
DMRTA1 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.64kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
ESGNESEWVKDLTSTKASLPTVSSRPRDPLDILTKIFPNYRRSRLEGILRFCKGDVVQAIEQVLNGKEHKPDNRNLANSEELENTAFQRA | |
RUO | |
DMRTA1 | |
Wheat Germ (in vitro) | |
GST |