Learn More
Abnova™ Human DMAP1 Partial ORF (NP_061973, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00055929-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a subunit of several, distinct complexes involved in the repression or activation of transcription. The encoded protein can independently repress transcription and is targeted to replication foci throughout S phase by interacting directly with the N-terminus of DNA methyltransferase 1. During late S phase, histone deacetylase 2 is added to this complex, providing a means to deacetylate histones in transcriptionally inactive heterochromatin following replication. The encoded protein is also a component of the nucleosome acetyltransferase of H4 complex and interacts with the transcriptional corepressor tumor susceptibility gene 101 and the pro-apoptotic death-associated protein 6, among others. Alternatively spliced transcript variants encoding the same protein have been described. [provided by RefSeq]
Sequence: MATGADVRDILELGGPEGDAASGTISKKDIINPDKKKSKKSSETLTFKRPEGMHREVYALLYSDKKDAPPLLPSDTGQGYRTVKAKLGSKKVRPWKWMPFEspecificaciones
NP_061973 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MATGADVRDILELGGPEGDAASGTISKKDIINPDKKKSKKSSETLTFKRPEGMHREVYALLYSDKKDAPPLLPSDTGQGYRTVKAKLGSKKVRPWKWMPF | |
RUO | |
DMAP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
55929 | |
DMAP1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686L09142/DNMAP1/DNMTAP1/EAF2/FLJ11543/KIAA1425/SWC4 | |
DMAP1 | |
Recombinant | |
wheat germ expression system |