missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human DKFZP781I1119 Full-length ORF (AAH41348.2, 1 a.a. - 522 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00166968-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: MLVHDYDDERTLEEEEMMDEGKNFSSEIEDLEKEGTMPLEDLLAFYGYEPTIPAVANSSANSSPSELADELPDMTLDKEEIAKDLLSGDDEETQSSADGLTPSVTSHETSDFFPRPLRSNTACDGDKESEVEDVETDSGNSPEDLRKEIMIGLQYQAEIPPYLGEYDGNEKVYENEDQLLWCPDVVLESKVKEYLVETSLRTGSEKIMDRISAGTHTRDNEQALYELLKCNHNIKEAIERYCCNGKASQGMTAWTEEECRSFEHALMLFGKDFHLIQKNKVRTRTVAECVAFYYMWKKSERYDYFAQQTRFGKKRYNHHPGVTDYMDRLVDETEALGGTVNASALTSNRPEPIPDQQLNILNSFTASDLTALTNSVATVCDPTDVNCLDDSFPPLGNTPRGQVNHVPVVTEELLTLPSNGESDCFNLFETGFYHSELNPMNMCSEESERPAKRLKMGIAVPESFMNEVSVNNLGVDFENHTHHITSAKMAVSVADFGSLSANETNGFISAHALHQHAALHSEEspecificaciones
AAH41348.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
84.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686L09111/DKFZp781G1119/DKFZp781I1119/FLJ35954 | |
MIER3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
166968 | |
DKFZP781I1119 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MLVHDYDDERTLEEEEMMDEGKNFSSEIEDLEKEGTMPLEDLLAFYGYEPTIPAVANSSANSSPSELADELPDMTLDKEEIAKDLLSGDDEETQSSADGLTPSVTSHETSDFFPRPLRSNTACDGDKESEVEDVETDSGNSPEDLRKEIMIGLQYQAEIPPYLGEYDGNEKVYENEDQLLWCPDVVLESKVKEYLVETSLRTGSEKIMDRISAGTHTRDNEQALYELLKCNHNIKEAIERYCCNGKASQGMTAWTEEECRSFEHALMLFGKDFHLIQKNKVRTRTVAECVAFYYMWKKSERYDYFAQQTRFGKKRYNHHPGVTDYMDRLVDETEALGGTVNASALTSNRPEPIPDQQLNILNSFTASDLTALTNSVATVCDPTDVNCLDDSFPPLGNTPRGQVNHVPVVTEELLTLPSNGESDCFNLFETGFYHSELNPMNMCSEESERPAKRLKMGIAVPESFMNEVSVNNLGVDFENHTHHITSAKMAVSVADFGSLSANETNGFISAHALHQHAALHSE | |
RUO | |
MIER3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |