Learn More
Abnova™ Human DEGS2 Full-length ORF (NP_996801.1, 1 a.a. - 323 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00123099-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a bifunctional enzyme that is involved in the biosynthesis of phytosphingolipids in human skin and in other phytosphingolipid-containing tissues. This enzyme can act as a sphingolipid delta(4)-desaturase, and also as a sphingolipid C4-hydroxylase. [provided by RefSeq]
Sequence: MGNSASRSDFEWVYTDQPHTQRRKEILAKYPAIKALMRPDPRLKWAVLVLVLVQMLTCWLVRGLAWRWLLFWAYAFGGCVNHSLTLAIHDISHNAAFGTGRAARNRWLAVFANLPVGVPYAASFKKYHVDHHRYLGGDGLDVDVPTRLEGWFFCTPARKLLWLVLQPFFYSLRPLCVHPKAVTRMEVLNTLVQLAADLAIFALWGLKPVVYLLASSFLGLGLHPISGHFVAEHYMFLKGHETYSYYGPLNWITFNVGYHVEHHDFPSIPGYNLPLVRKIAPEYYDHLPQHHSWVKVLWDFVFEDSLGPYARVKRVYRLAKDGLEspecificaciones
NP_996801.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
63.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C14orf66/DES2/FADS8 | |
DEGS2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
123099 | |
DEGS2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGNSASRSDFEWVYTDQPHTQRRKEILAKYPAIKALMRPDPRLKWAVLVLVLVQMLTCWLVRGLAWRWLLFWAYAFGGCVNHSLTLAIHDISHNAAFGTGRAARNRWLAVFANLPVGVPYAASFKKYHVDHHRYLGGDGLDVDVPTRLEGWFFCTPARKLLWLVLQPFFYSLRPLCVHPKAVTRMEVLNTLVQLAADLAIFALWGLKPVVYLLASSFLGLGLHPISGHFVAEHYMFLKGHETYSYYGPLNWITFNVGYHVEHHDFPSIPGYNLPLVRKIAPEYYDHLPQHHSWVKVLWDFVFEDSLGPYARVKRVYRLAKDGL | |
RUO | |
DEGS2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |