Learn More
Abnova™ Human DEGS1 Partial ORF (NP_003667, 226 a.a. - 323 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00008560-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a member of the membrane fatty acid desaturase family which is responsible for inserting double bonds into specific positions in fatty acids. This protein contains three His-containing consensus motifs that are characteristic of a group of membrane fatty acid desaturases. It is predicted to be a multiple membrane-spanning protein localized to the endoplasmic reticulum. Overexpression of this gene inhibited biosynthesis of the EGF receptor, suggesting a possible role of a fatty acid desaturase in regulating biosynthetic processing of the EGF receptor. Two splice variants have been identified. [provided by RefSeq]
Sequence: SGHFIAEHYMFLKGHETYSYYGPLNLLTFNVGYHNEHHDFPNIPGKSLPLVRKIAAEYYDNLPHYNSWIKVLYDFVMDDTISPYSRMKRHQKGEMVLEEspecificaciones
NP_003667 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SGHFIAEHYMFLKGHETYSYYGPLNLLTFNVGYHNEHHDFPNIPGKSLPLVRKIAAEYYDNLPHYNSWIKVLYDFVMDDTISPYSRMKRHQKGEMVLE | |
RUO | |
DEGS1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8560 | |
DEGS1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DEGS/DES1/Des-1/FADS7/MGC5079/MIG15/MLD | |
DEGS1 | |
Recombinant | |
wheat germ expression system |