missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DDX26 Partial ORF (NP_036273, 779 a.a. - 887 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00026512-Q01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16198342

  • 335.00€ / 10 microgramos
Fecha estimada de envío: 10-06-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. The protein encoded by this gene is a DEAD box protein that is part of a complex that interacts with the C-terminus of RNA polymerase II and is involved in 3' end processing of snRNAs. In addition, this gene is a candidate tumor suppressor and located in the critical region of loss of heterozygosity (LOH). Three transcript variants encoding two different isoforms have been found for this gene. [provided by RefSeq]

Sequence: DKEQCAEENIPASSLNKGKKLMHCRSHEEVNTELKAQIMKEIRKPGRKYERIFTLLKHVQGSLQTRLIFLQNVIKEASRFKKRMLIEQLENFLDEIHRRANQINHINSN
Especificaciones

Especificaciones

NP_036273
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.73kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
DKEQCAEENIPASSLNKGKKLMHCRSHEEVNTELKAQIMKEIRKPGRKYERIFTLLKHVQGSLQTRLIFLQNVIKEASRFKKRMLIEQLENFLDEIHRRANQINHINSN
RUO
INTS6
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
26512
DDX26 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DBI-1/DDX26/DDX26A/DICE1/DKFZp434B105/HDB/INT6/Notchl2
INTS6
Recombinant
wheat germ expression system
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales