Learn More
Abnova™ Human DDX26 Partial ORF (NP_036273, 779 a.a. - 887 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00026512-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. The protein encoded by this gene is a DEAD box protein that is part of a complex that interacts with the C-terminus of RNA polymerase II and is involved in 3' end processing of snRNAs. In addition, this gene is a candidate tumor suppressor and located in the critical region of loss of heterozygosity (LOH). Three transcript variants encoding two different isoforms have been found for this gene. [provided by RefSeq]
Sequence: DKEQCAEENIPASSLNKGKKLMHCRSHEEVNTELKAQIMKEIRKPGRKYERIFTLLKHVQGSLQTRLIFLQNVIKEASRFKKRMLIEQLENFLDEIHRRANQINHINSNEspecificaciones
NP_036273 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DKEQCAEENIPASSLNKGKKLMHCRSHEEVNTELKAQIMKEIRKPGRKYERIFTLLKHVQGSLQTRLIFLQNVIKEASRFKKRMLIEQLENFLDEIHRRANQINHINSN | |
RUO | |
INTS6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
26512 | |
DDX26 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DBI-1/DDX26/DDX26A/DICE1/DKFZp434B105/HDB/INT6/Notchl2 | |
INTS6 | |
Recombinant | |
wheat germ expression system |