missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DAZ4 Full-length ORF (NP_065153.1, 1 a.a. - 390 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00057135-P01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16172523

  • 349.00€ / 10 microgramos
Fecha estimada de envío: 31-10-2023
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene is a member of the DAZ gene family and is a candidate for the human Y-chromosomal azoospermia factor (AZF). Its expression is restricted to premeiotic germ cells, particularly in spermatogonia. It encodes an RNA-binding protein that is important for spermatogenesis. Four copies of this gene are found on chromosome Y within palindromic duplications; one pair of genes is part of the P2 palindrome and the second pair is part of the P1 palindrome. Each gene contains a 2.4 kb repeat including a 72-bp exon, called the DAZ repeat; the number of DAZ repeats is variable and there are several variations in the sequence of the DAZ repeat. Each copy of the gene also contains a 10.8 kb region that may be amplified; this region includes five exons that encode an RNA recognition motif (RRM) domain. This gene contains two copies of the 10.8 kb repeat. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq]

Sequence: MSAANPETPNSTISREASTQSSSAAASQGWVLPEGKIVPNTVFVGGIDARMDETEIGSCFGRYGSVKEVKIITNRTGVSKGYGFVSFVNDVDVQKIVGSQIHFHGKKLKLGPAIRKQKLCARHVQPRPLVVNPPPPPQFQNVWRNPNTETYLQPQITPNPVTQHVQAYSAYPHSPGQVITGCQLLVYNYQEYPTYPDSAFQVTTGYQLPVYNYQPFPAYPRSPFQVTAGYQLPVYNYQAFPAYPNSPFQVATGYQFPVYNYQPFPAYPSSPFQVTAGYQLPVYNYQAFPAYPNSPFQVATGYQFPVYNYQAFPAYPNSPVQVTTGYQLPVYNYQAFPAYPNSAVQVTTGYQFHVYNYQMPPQCPVGEQRRNLWTEAYKWWYLVCLIQRRD
Especificaciones

Especificaciones

NP_065153.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
70.5kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DAZ/DAZ1/pDP1680/pDP1681
DAZ4
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
57135
DAZ4 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSAANPETPNSTISREASTQSSSAAASQGWVLPEGKIVPNTVFVGGIDARMDETEIGSCFGRYGSVKEVKIITNRTGVSKGYGFVSFVNDVDVQKIVGSQIHFHGKKLKLGPAIRKQKLCARHVQPRPLVVNPPPPPQFQNVWRNPNTETYLQPQITPNPVTQHVQAYSAYPHSPGQVITGCQLLVYNYQEYPTYPDSAFQVTTGYQLPVYNYQPFPAYPRSPFQVTAGYQLPVYNYQAFPAYPNSPFQVATGYQFPVYNYQPFPAYPSSPFQVTAGYQLPVYNYQAFPAYPNSPFQVATGYQFPVYNYQAFPAYPNSPVQVTTGYQLPVYNYQAFPAYPNSAVQVTTGYQFHVYNYQMPPQCPVGEQRRNLWTEAYKWWYLVCLIQRRD
RUO
DAZ4
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales