Learn More
ACROBiosystems Human / Cynomolgus / Rhesus macaque CD28 Protein, Fc Tag
HPLC-verified
Marca:  ACROBiosystems CD8-H525a-100ug
Detalles adicionales : Peso : 0.00010kg
 Ver más versiones de este producto
Código de producto. 16477329
  • 201.17€ / 100 microgramos
Fecha estimada de envío: 21-07-2022
para ver el stock
Descripción
Descripción
T-cell-specific surface glycoprotein CD28 is also known as TP44, is a single-pass type I membrane protein which contains one Ig-like V-type (immunoglobulin-like) domain. is one of the molecules expressed on T cells that provide co-stimulatory signals, which are required for T cell activation. CD28 is the receptor for CD80 (B7.1) and CD86 (B7.2). When activated by Toll-like receptor ligands, the CD80 expression is upregulated in antigen presenting cells (APCs). The CD86 expression on antigen presenting cells is constitutive. CD28 is the only B7 receptor constitutively expressed on naive T cells.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 41.6 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Especificaciones
Especificaciones
NP_006130.1
41.6 kDa
100 µg
Asn 19 - Pro 152
CD28,Tp44
Unconjugated
Tris with Glycine, Arginine and NaCl, pH7.5
HEK293
Human / Cynomolgus / Rhesus macaque
Powder
>95% as determined by SDS-PAGE
Human / Cynomolgus / Rhesus macaque CD28, Fc Tag (HPLC-verified) (CD8-H525a) is expressed from human 293 cells (HEK293). It contains AA Asn 19 - Pro 152 (Accession # P10747-1). In the region Asn 19 - Pro 152, the AA sequence of Human, Cynomolgus and Rhesus macaque CD28 are homologus.
-20°C
CD28
Fc Tag
Less than 1.0 EU / µg by the LAL method.
Immune Checkpoint Proteins