missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CRTC1 Partial ORF (NP_056136.1, 553 a.a. - 634 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00023373-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: ASHSGIPNIILTVTGESPPSLSKELTSSLAGVGDVSFDSDSQFPLDELKIDPLTLDGLHMLNDPDMVLADPATEDTFRMDRLEspecificaciones
NP_056136.1 | |
Liquid | |
23373 | |
CRTC1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ14027/KIAA0616/MECT1/TORC1/WAMTP1 | |
CRTC1 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.76kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
ASHSGIPNIILTVTGESPPSLSKELTSSLAGVGDVSFDSDSQFPLDELKIDPLTLDGLHMLNDPDMVLADPATEDTFRMDRL | |
RUO | |
CRTC1 | |
Wheat Germ (in vitro) | |
GST |