Learn More
Abnova™ Human COPS3 Full-length ORF (NP_003644.2, 1 a.a. - 423 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00008533-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene possesses kinase activity that phosphorylates regulators involved in signal transduction. It phosphorylates I kappa-Balpha, p105, and c-Jun. It acts as a docking site for complex-mediated phosphorylation. The gene is located within the Smith-Magenis syndrome region on chromosome 17. [provided by RefSeq]
Sequence: MASALEQFVNSVRQLSAQGQMTQLCELINKSGELLAKNLSHLDTVLGALDVQEHSLGVLAVLFVKFSMPSVPDFETLFSQVQLFISTCNGEHIRYATDTFAGLCHQLTNALVERKQPLRGIGILKQAIDKMQMNTNQLTSIHADLCQLCLLAKCFKPALPYLDVDMMDICKENGAYDAKHFLCYYYYGGMIYTGLKNFERALYFYEQAITTPAMAVSHIMLESYKKYILVSLILLGKVQQLPKYTSQIVGRFIKPLSNAYHELAQVYSTNNPSELRNLVNKHSETFTRDNNMGLVKQCLSSLYKKNIQRLTKTFLTLSLQDMASRVQLSGPQEAEKYVLHMIEDGEIFASINQKDGMVSFHDNPEKYNNPAMLHNIDQEMLKCIELDERLKAMDQEITVNPQFVQKSMGSQEDDSGNKPSSYSEspecificaciones
NP_003644.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
74.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CSN3/SGN3 | |
COPS3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
8533 | |
COPS3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MASALEQFVNSVRQLSAQGQMTQLCELINKSGELLAKNLSHLDTVLGALDVQEHSLGVLAVLFVKFSMPSVPDFETLFSQVQLFISTCNGEHIRYATDTFAGLCHQLTNALVERKQPLRGIGILKQAIDKMQMNTNQLTSIHADLCQLCLLAKCFKPALPYLDVDMMDICKENGAYDAKHFLCYYYYGGMIYTGLKNFERALYFYEQAITTPAMAVSHIMLESYKKYILVSLILLGKVQQLPKYTSQIVGRFIKPLSNAYHELAQVYSTNNPSELRNLVNKHSETFTRDNNMGLVKQCLSSLYKKNIQRLTKTFLTLSLQDMASRVQLSGPQEAEKYVLHMIEDGEIFASINQKDGMVSFHDNPEKYNNPAMLHNIDQEMLKCIELDERLKAMDQEITVNPQFVQKSMGSQEDDSGNKPSSYS | |
RUO | |
COPS3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |