missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human COL8A2 Partial ORF (NP_005193.1, 626 a.a. - 696 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00001296-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: VHVKGTNVWVALYKNNVPATYTYDEYKKGYLDQASGGAVLQLRPNDQVWVQMPSDQANGLYSTEYIHSSFSEspecificaciones
NP_005193.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.55kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VHVKGTNVWVALYKNNVPATYTYDEYKKGYLDQASGGAVLQLRPNDQVWVQMPSDQANGLYSTEYIHSSFS | |
RUO | |
COL8A2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
1296 | |
COL8A2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FECD/FLJ00201/MGC116970/MGC116972/PPCD/PPCD2 | |
COL8A2 | |
Recombinant | |
wheat germ expression system |