missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CHRNA2 Partial ORF (NP_000733, 27 a.a. - 136 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00001135-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: EEAKRPPPRAPGDPLSSPSPTALPQGGSHTETEDRLFKHLFRGYNRWARPVPNTSDVVIVRFGLSIAQLIDVDEKNQMMTTNVWLKQEWSDYKLRWNPADFGNITSLRVPEspecificaciones
NP_000733 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
EEAKRPPPRAPGDPLSSPSPTALPQGGSHTETEDRLFKHLFRGYNRWARPVPNTSDVVIVRFGLSIAQLIDVDEKNQMMTTNVWLKQEWSDYKLRWNPADFGNITSLRVP | |
RUO | |
CHRNA2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
1135 | |
CHRNA2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CHRNA2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |