Learn More
Abnova™ Human CHGB Full-length ORF (NP_001810.1, 1 a.a. - 677 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00001114-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a tyrosine-sulfated secretory protein abundant in peptidergic endocrine cells and neurons. This protein may serve as a precursor for regulatory peptides. [provided by RefSeq]
Sequence: MQPTLLLSLLGAVGLAAVNSMPVDNRNHNEGMVTRCIIEVLSNALSKSSAPPITPECRQVLKTSRKDVKDKETTENENTKFEVRLLRDPADASEAHESSSRGEAGAPGEEDIQGPTKADTEKWAEGGGHSRERADEPQWSLYPSDSQVSEEVKTRHSEKSQREDEEEEEGENYQKGERGEDSSEEKHLEEPGETQNAFLNERKQASAIKKEELVARSETHAAGHSQEKTHSREKSSQESGEEAGSQENHPQESKGQPRSQEESEEGEEDATSEVDKRRTRPRHHHGRSRPDRSSQGGSLPSEEKGHPQEESEESNVSMASLGEKRDHHSTHYRASEEEPEYGEEIKGYPGVQAPEDLEWERYRGRGSEEYRAPRPQSEESWDEEDKRNYPSLELDKMAHGYGEESEEERGLEPGKGRHHRGRGGEPRAYFMSDTREEKRFLGEGHHRVQENQMDKARRHPQGAWKELDRNYLNYGEEGAPGKWQQQGDLQDTKENREEARFQDKQYSSHHTAEKRKRLGELFNPYYDPLQWKSSHFERRDNMNDNFLEGEEENELTLNEKNFFPEYNYDWWEKKPFSEDVNWGYEKRNLARVPKLDLKRQYDRVAQLDQLLHYRKKSAEFPDFYDSEEPVSTHQEAENEKDRADQTVLTEDEKKELENLAAMDLELQKIAEKFSQRGEspecificaciones
NP_001810.1 | |
Liquid | |
1114 | |
CHGB (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MQPTLLLSLLGAVGLAAVNSMPVDNRNHNEGMVTRCIIEVLSNALSKSSAPPITPECRQVLKTSRKDVKDKETTENENTKFEVRLLRDPADASEAHESSSRGEAGAPGEEDIQGPTKADTEKWAEGGGHSRERADEPQWSLYPSDSQVSEEVKTRHSEKSQREDEEEEEGENYQKGERGEDSSEEKHLEEPGETQNAFLNERKQASAIKKEELVARSETHAAGHSQEKTHSREKSSQESGEEAGSQENHPQESKGQPRSQEESEEGEEDATSEVDKRRTRPRHHHGRSRPDRSSQGGSLPSEEKGHPQEESEESNVSMASLGEKRDHHSTHYRASEEEPEYGEEIKGYPGVQAPEDLEWERYRGRGSEEYRAPRPQSEESWDEEDKRNYPSLELDKMAHGYGEESEEERGLEPGKGRHHRGRGGEPRAYFMSDTREEKRFLGEGHHRVQENQMDKARRHPQGAWKELDRNYLNYGEEGAPGKWQQQGDLQDTKENREEARFQDKQYSSHHTAEKRKRLGELFNPYYDPLQWKSSHFERRDNMNDNFLEGEEENELTLNEKNFFPEYNYDWWEKKPFSEDVNWGYEKRNLARVPKLDLKRQYDRVAQLDQLLHYRKKSAEFPDFYDSEEPVSTHQEAENEKDRADQTVLTEDEKKELENLAAMDLELQKIAEKFSQRG | |
RUO | |
CHGB | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
104.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SCG1 | |
CHGB | |
Yes | |
wheat germ expression system |