Learn More
ACROBiosystems Human CG alpha / CGA Protein, His Tag
Marca: ACROBiosystems CGA-H52H2-1mg
Detalles adicionales : Peso : 0.00010kg
Descripción
Chorionic gonadotrophin subunit alpha (CGA),a member of the glycoprotein hormones subunit alpha family, is also known as glycoprotein hormones alpha chain, choriogonadotropin alpha chain, follicle-stimulating hormone alpha chain (FSH-alpha), luteinizing hormone alpha chain (LSH-alpha) and thyroid-stimulating hormone alpha chain (TSH-alpha). The hormones human chorionic gonadotropin (hCG), luteinizing hormone (LH), follicle-stimulating hormone (FSH), and thyroid-stimulating hormone (TSH) are dimers consisting of alpha and beta subunits that are associated noncovalently. The alpha subunits (CGA) of these four human glycoprotein hormones are nearly identical, however, their beta chains are unique and confer biological specificity.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 12.1 kDa. The protein migrates as 22-26 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
AAH20782 | |
1 mg | |
Ala 25 - Ser 116 | |
CGA,TSH-alpha,LSH-alpha,CG-alpha,FSH-alpha | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>92% as determined by SDS-PAGE |
12.1 kDa | |
Human CG alpha, His Tag (CGA-H52H2) is expressed from human 293 cells (HEK293). It contains AA Ala 25 - Ser 116 (Accession # AAH20782). | |
-20°C | |
CG alpha | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Cytokines and Growth Factors |