missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CFLAR Partial ORF (AAH01602, 178 a.a. - 277 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00008837-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Description
Sequence: AGTSYRNVLQAAIQKSLKDPSNNFRLHNGRSKEQRLKEQLGAQQEPVKKSIQESEAFLPQSIPEERYKMKSKPLGICLIIDCIGNETELLRDTFTSLGYESpecifications
AAH01602 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
AGTSYRNVLQAAIQKSLKDPSNNFRLHNGRSKEQRLKEQLGAQQEPVKKSIQESEAFLPQSIPEERYKMKSKPLGICLIIDCIGNETELLRDTFTSLGYE | |
RUO | |
CFLAR | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8837 | |
CFLAR (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CASH/CASP8AP1/CLARP/Casper/FLAME/FLAME-1/FLAME1/FLIP/I-FLICE/MRIT/c-FLIP/c-FLIPL/c-FLIPR/c-FLIPS | |
CFLAR | |
Recombinant | |
wheat germ expression system |