Learn More
Abnova™ Human CFL2 Partial ORF (NP_068733, 57 a.a. - 166 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00001073-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes an intracellular protein that is involved in the regulation of actin-filament dynamics. This protein is a major component of intranuclear and cytoplasmic actin rods. It can bind G- and F-actin in a 1:1 ratio of cofilin to actin, and it reversibly controls actin polymerization and depolymerization in a pH-dependent manner. Mutations in this gene cause nemaline myopathy type 7, a form of congenital myopathy. Alternative splicing results in multiple transcript variants. [provided by RefSeq]
Sequence: VGDIGDTVEDPYTSFVKLLPLNDCRYALYDATYETKESKKEDLVFIFWAPESAPLKSKMIYASSKDAIKKKFTGIKHEWQVNGLDDIKDRSTLGEKLGGNVVVSLEGKPLEspecificaciones
NP_068733 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VGDIGDTVEDPYTSFVKLLPLNDCRYALYDATYETKESKKEDLVFIFWAPESAPLKSKMIYASSKDAIKKKFTGIKHEWQVNGLDDIKDRSTLGEKLGGNVVVSLEGKPL | |
RUO | |
CFL2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
1073 | |
CFL2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NEM7 | |
CFL2 | |
Recombinant | |
wheat germ expression system |