Learn More
Abnova™ Human CDKL5 Partial ORF (AAH36091, 722 a.a. - 831 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00006792-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene is a member of Ser/Thr protein kinase family and encodes a phosphorylated protein with protein kinase activity. Mutations in this gene have been associated with X-linked infantile spasm syndrome (ISSX), also known as X-linked West syndrome, and Rett syndrome (RTT). Alternate transcriptional splice variants have been characterized. [provided by RefSeq]
Sequence: RPDNSFHENNVSTRVSSLPSESSSGTNHSKRQPAFDPWKSPENISHSEQLKEKEKQGFFRSMKKKKKKSQTVPNSDSPDLLTLQKSIHSASTPSSRPKEWRPEKISDLQTEspecificaciones
AAH36091 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RPDNSFHENNVSTRVSSLPSESSSGTNHSKRQPAFDPWKSPENISHSEQLKEKEKQGFFRSMKKKKKKSQTVPNSDSPDLLTLQKSIHSASTPSSRPKEWRPEKISDLQT | |
RUO | |
CDKL5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
6792 | |
CDKL5 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ISSX/STK9 | |
CDKL5 | |
Recombinant | |
wheat germ expression system |