Learn More
ACROBiosystems Human CD8 alpha&beta (CD8A&CD8B) Heterodimer Protein, His Tag&Tag Free
MALS verified
Marca:  ACROBiosystems CDA-H52W5-1mg
Detalles adicionales : Peso : 0.00010kg
 Ver más versiones de este producto
Código de producto. 17186181
  • 3512.64€ / 1 miligramo
Fecha estimada de envío: 05-01-2023
para ver el stock
Descripción
Descripción
Integral membrane glycoprotein that plays an essential role in the immune response and serves multiple functions in responses against both external and internal offenses. CD8A forms disulfide-linked heterodimers with CD8B at the cell surface. CD8A homodimer molecules also promote the survival and differentiation of activated lymphocytes into memory CD8 T-cells. Additionally, CD8B plays a critical role in thymic selection of CD8+ T-cells. A palmitoylation site in the cytoplasmic tail of CD8B chain contributes to partitioning of CD8 into the plasma membrane lipid rafts where signaling proteins are enriched.
Human CD8A&CD8B Heterodimer Protein, His Tag&Tag Free, produced by co-expression of CD8 alpha and CD8 beta, has a calculated MW of 19.5 kDa (CD8 alpha) and 22.1 kDa (CD8 beta). Subunit CD8 alpha is fused with a polyhistidine tag at the C-terminus and subu
Especificaciones
Especificaciones
NP_001759.3 (CD8A)/NP_004922.1 (CD8B)
19.5 kDa (CD8A) and 22.1 kDa (CD8B)
1 mg
Ser 22 - Asp 182 (CD8A) and Leu 22 - Pro 170 (CD8B)
CD8 alpha and beta,CD8A and CD8B
Unconjugated
PBS, pH7.4
HEK293
Human
Powder
>95% as determined by SDS-PAGE
Human CD8A&CD8B; Heterodimer Protein, His Tag&Tag; Free (CDA-H52W5) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Asp 182 (CD8A) and Leu 22 - Pro 170 (CD8B) (Accession # P01732-1 (CD8A) and P10966-1 (CD8B)).
-20°C
CD8 alpha and beta
His Tag
Less than 1.0 EU / µg by the LAL method.
Bio-Markers and CD Antigens