missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CD5 Partial ORF (ABM83003, 403 a.a. - 495 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00000921-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: KKLVKKFRQKKQRQWIGPTGMNQNMSFHRNHTATVRSHAENPTASHVDNEYSQPPRNSRLSAYPALEGVLHRSSMQPDNSSDSDYDLHGAQRLEspecificaciones
ABM83003 | |
Liquid | |
921 | |
CD5 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
LEU1/T1 | |
CD5 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.86kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
KKLVKKFRQKKQRQWIGPTGMNQNMSFHRNHTATVRSHAENPTASHVDNEYSQPPRNSRLSAYPALEGVLHRSSMQPDNSSDSDYDLHGAQRL | |
RUO | |
CD5 | |
Wheat Germ (in vitro) | |
GST |