Learn More
ACROBiosystems Human CD40 / TNFRSF5 Protein, Mouse IgG2a Fc Tag, low endotoxin
MALS verified
Marca:  ACROBiosystems CD0-H525A-1MG
Detalles adicionales : Peso : 0.00001kg
 Ver más versiones de este producto
Código de producto. 15896127
  • 2639.24€ / 1 miligramo
Fecha estimada de envío: 11-04-2024
para ver el stock
Descripción
Descripción
CD40 is also known as TNFRSF5, Bp50, CDW40, MGC9013, TNFRSF5 and p50, is a member of the TNF receptor superfamily which are single transmembrane-spanning glycoproteins, and plays an essential role in mediating a broad variety of immune and inflammatory responses including T cell-dependent immunoglobulin class switching, memory B cell development, and germinal center formation. CD40 is a costimulatory protein found on antigen presenting cells and is required for their activation. The binding of CD154 (CD40L) on TH cells to CD40 activates antigen presenting cells and induces a variety of downstream effects. CD40 contains 4 cysteine-rich repeats in the extracellular domain, and is expressed in B cells, dendritic cells, macrophages, endothelial cells, and several tumor cell lines.
This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 46.1 kDa. The protein migrates as 58 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Especificaciones
Especificaciones
NP_001241.1
1 mg
Glu 21 - Arg 193
CD40,Bp50,CDW40,MGC9013,TNFRSF5,p50
Unconjugated
Mouse IgG Fc Tag
Less than 0.1 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
46.1 kDa
Human CD40, Mouse IgG2a Fc Tag, low endotoxin (MALS verified) (CD0-H525a) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Arg 193 (Accession # P25942-1).
-20°C
CD40
Human
50 mM Tris, 100 mM Glycine, 150 mM NaCl, pH7.5
HEK293
Immune Checkpoint Proteins