Learn More
ACROBiosystems Human CD300LG / Nepmucin Protein, His Tag
Marca: ACROBiosystems CDG-H5223-100UG
Detalles adicionales : Peso : 0.00001kg
Descripción
CMRF35-like molecule 9 (CLM-9) is also known as CD300 antigen-like family member G (CD300LG or CD300g), Nepmucin, Triggering receptor expressed on myeloid cells 4 (TREM-4), which belongs to the CD300 family. CD300LG contains one Ig-like V-type (immunoglobulin-like) domain. CD300LG is highly expressed in heart, skeletal muscle and placenta. CD300LG is a receptor which may mediate L-selectin-dependent lymphocyte rollings. CD300LG binds SELL in a calcium dependent manner and also binds lymphocyte.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.6 kDa. The protein migrates as 33-68 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.Especificaciones
AAH25395 | |
100 μg | |
Leu 19 - Arg 247 | |
CD300LG,CLM9,TREM4,CD300g,Nepmucin | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
25.6 kDa | |
Human CD300LG, His Tag (CDG-H5223) is expressed from human 293 cells (HEK293). It contains AA Leu 19 - Arg 247 (Accession # AAH25395). | |
-20°C | |
CD300LG | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |