missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CD300A Full-length ORF (NP_009192.2, 1 a.a. - 299 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00011314-P01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16166512

  • 349.00€ / 10 microgramos
Fecha estimada de envío: 30-10-2023
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The CMRF35 antigen (CMRF35A; MIM 606786), which was identified by reactivity with a monoclonal antibody, is present on monocytes, neutrophils, and some T and B lymphocytes. CMRF35H is recognized by the same antibody and is distinct from CMRF35 (Green et al., 1998 [PubMed 9701027]).[supplied by OMIM]

Sequence: MWLPWALLLLWVPGCFALSKCRTVAGPVGGSLSVQCPYEKEHRTLNKYWCRPPQIFLCDKIVETKGSAGKRNGRVSIRDSPANLSFTVTLENLTEEDAGTYWCGVDTPWLRDFHDPVVEVEVSVFPASTSMTPASITAAKTSTITTAFPPVSSTTLFAVGATHSASIQEETEEVVNSQLPLLLSLLALLLLLLVGASLLAWRMFQKWIKAGDHSELSQNPKQAATQSELHYANLELLMWPLQEKPAPPREVEVEYSTVASPREELHYASVVFDSNTNRIAAQRPREEEPDSDYSVIRKT
Especificaciones

Especificaciones

NP_009192.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
59.6kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CMRF-35-H9/CMRF-35H/CMRF35H/CMRF35H9/IGSF12/IRC1/IRC2/IRp60
CD300A
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
11314
CD300A (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MWLPWALLLLWVPGCFALSKCRTVAGPVGGSLSVQCPYEKEHRTLNKYWCRPPQIFLCDKIVETKGSAGKRNGRVSIRDSPANLSFTVTLENLTEEDAGTYWCGVDTPWLRDFHDPVVEVEVSVFPASTSMTPASITAAKTSTITTAFPPVSSTTLFAVGATHSASIQEETEEVVNSQLPLLLSLLALLLLLLVGASLLAWRMFQKWIKAGDHSELSQNPKQAATQSELHYANLELLMWPLQEKPAPPREVEVEYSTVASPREELHYASVVFDSNTNRIAAQRPREEEPDSDYSVIRKT
RUO
CD300A
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales