Learn More
ACROBiosystems Human CD27 Ligand / CD70 Protein, Mouse IgG2a Fc Tag, low endotoxin
low endotoxin
Marca: ACROBiosystems CDL-H525A-1MG
Detalles adicionales : Peso : 0.00001kg
Descripción
Cluster of Differentiation 70 (CD70) is also known as CD27 ligand (CD27L / CD27LG), TNFSF7, TNFSF7G, is a type II transmembrane glycoprotein belonging to the TNF superfamily (TNFSF) and is a surface antigen found on activated T-and B-lymphocytes and mature dendritic cells. Binding of CD70 to its receptor CD27 induces in priming, effector functions, differentiation and memory formation of T-cells, and thus is invloved in the biological processes including T-cell activation, the proliferation of costimulated T-cells, as well as the generation of cytolytic T-cells. CD70 on T cells provides costimulatory signals that are required for T cell proliferation, clonal expansion and the promotion of effector T cell formation. CD70 on mouse B cell has been shown to inhibit terminal differentiation of activated B cells into plasma cells and enhances commitment to memory B cell responses. CD70 induces proliferation and IFNγ production, on NK cells.
This protein carries a mouse IgG2a Fc tag at the N-terminus. The protein has a calculated MW of 44.0 kDa. The protein migrates as 50-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
NP_001243.1 | |
44.0 kDa | |
1 mg | |
Gln 39 - Pro 193 | |
CD70,CD27LG,TNFSF7,TNFSF7G,CD27L | |
Unconjugated | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Human |
Powder | |
>85% as determined by SDS-PAGE | |
Human CD27 Ligand, Mouse IgG2a Fc Tag, low endotoxin (CDL-H525a) is expressed from human 293 cells (HEK293). It contains AA Gln 39 - Pro 193 (Accession # P32970-1). | |
-20°C | |
CD27 Ligand | |
Mouse IgG Fc Tag | |
Less than 0.1 EU / µg by the LAL method. | |
Immune Checkpoint Proteins |