Learn More
Abnova™ Human CARD6 Partial ORF (NP_115976, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00084674-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a protein that contains a caspase recruitment domain (CARD), an antiparallel six-helical bundle that mediates homotypic protein-protein interactions. The encoded protein is a microtubule-associated protein that has been shown to interact with receptor-interacting protein kinases and positively modulate signal transduction pathways converging on activation of the inducible transcription factor NF-kB. [provided by RefSeq]
Sequence: MATESTPSEIIERERKKLLEILQHDPDSILDTLTSRRLISEEEYETLENVTDLLKKSRKLLILVQKKGEATCQHFLKCLFSTFPQSAAICGLRHEVLKHENTVPPQSMGAEspecificaciones
NP_115976 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MATESTPSEIIERERKKLLEILQHDPDSILDTLTSRRLISEEEYETLENVTDLLKKSRKLLILVQKKGEATCQHFLKCLFSTFPQSAAICGLRHEVLKHENTVPPQSMGA | |
RUO | |
CARD6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
84674 | |
CARD6 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CINCIN1 | |
CARD6 | |
Recombinant | |
wheat germ expression system |