Learn More
Abnova™ Human C4Orf49 Full-Length Orf (Aai04174.1, 1 A.A.-240 A.A.) Recombinant Protein With Gst Tag At N-Terminal.
Recombinant proteins expressed from in vitro wheat germ system. Such proteins mirror the conformation and folding of the native proteins in the eukaryotic biological system.
Marca: Abnova™ H00084709-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Good Folding
- Excellent amino acids labeling
Immunogen Sequence: MYLRRAVSKTLALPLRAPPNPAPLGKDASLRRMSSNRFPGSSGSNMIYYLVVGVTVSAGGYYAYKTVTSDQAKHTEHKTNLKEKTKAEIHPFQGEKENVAETEKASSEAPEELIVEAEVVDAEESPSATVVVIKEASACPGHVEAA
Especificaciones
AAI04174.1 | |
Liquid | |
84709 | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC125827/MGC125828/OSAP | |
C4orf49 |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH=8.0 in the elution buffer. | |
C4orf49 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
MYLRRAVSKTLALPLRAPPNPAPLGKDASLRRMSSNRFPGSSGSNMIYYLVVGVTVSAGGYYAYKTVTSDQAKHTEHKTNLKEKTKAEIHPFQGEKENVAETEKASSEAPEELIVEAEVVDAEESPSATVVVIKEASACPGHVEAAPETTAVSAETGPEVTDAAARETTEVNPETTPEVTNAALDEAVTIDNDKDTTKNETSDEYAELEEENSPAESESSAGDDLQEEASVGSEAASAQG | |
RUO | |
C4orf49 | |
GST |
For Research Use Only