missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human C10orf117 Partial ORF (NP_071896, 702 a.a. - 800 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00064318-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: PIVQRFAAHLIAGAPSEGSGALKPELSRRSATELFEAYSMAEMTFNPPVESSNPKIKGKFLQGDSFLNEDLNQLIKRYSSEVATESPLDFTKYLKTSLHEspecificaciones
NP_071896 | |
Liquid | |
64318 | |
C10orf117 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AD24/C10orf117/FAD24/FLJ12820 | |
NOC3L | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PIVQRFAAHLIAGAPSEGSGALKPELSRRSATELFEAYSMAEMTFNPPVESSNPKIKGKFLQGDSFLNEDLNQLIKRYSSEVATESPLDFTKYLKTSLH | |
RUO | |
NOC3L | |
Wheat Germ (in vitro) | |
GST |