missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human BRP44 Full-length ORF (NP_056230.1, 1 a.a. - 127 a.a.) Recombinant Protein MW: 40.7kDa with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00025874-P02.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: MSAAGARGLRATYHRLLDKVELMLPEKLRPLYNHPAGPRTVFFWAPIMKWGLVCAGLADMARPAEKLSTAQSAVLMATGFIWSRYSLVIIPKNWSLFAVNFFVGAAGASQLFRIWRYNQELKAKAHKEspecificaciones
NP_056230.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
40.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
BRP44 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
25874 | |
BRP44 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp564B167/MGC125752/MGC125753 | |
BRP44 | |
Recombinant | |
wheat germ expression system |