Learn More
Abnova™ Human BMF Full-length ORF (AAH69505.1, 1 a.a. - 184 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00090427-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene belongs to the BCL2 protein family. BCL2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. This protein contains a single BCL2 homology domain 3 (BH3), and has been shown to bind BCL2 proteins and function as an apoptotic activator. This protein is found to be sequestered to myosin V motors by its association with dynein light chain 2, which may be important for sensing intracellular damage and triggering apoptosis. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]
Sequence: MEPSQCVEELEDDVFQPEDGEPVTQPGSLLSADLFAQSLLDCPLSRLQLFPLTHCCGPGLRPTSQEDKATQTLSPASPSPGVMLPCGVTEEPQRLFYGNAGYRLPLPASFPAVLPIGEQPPEGQWQHQAEVQIARKLQCIADQFHRLHVQQHQQNQNRVWWQILLFLHNLALNGEENRNGAGPREspecificaciones
AAH69505.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
46.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
BMF | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
90427 | |
BMF (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ00065 | |
BMF | |
Recombinant | |
wheat germ expression system |