missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human BAI3 Partial ORF (NP_001695, 26 a.a. - 135 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00000577-Q01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16102131

  • 335.00€ / 10 microgramos
Fecha estimada de envío: 11-06-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This p53-target gene encodes a brain-specific angiogenesis inhibitor, a seven-span transmembrane protein, and is thought to be a member of the secretin receptor family. Brain-specific angiogenesis proteins BAI2 and BAI3 are similar to BAI1 in structure, have similar tissue specificities, and may also play a role in angiogenesis. [provided by RefSeq]

Sequence: QDFWCSTLVKGVIYGSYSVSEMFPKNFTNCTWTLENPDPTKYSIYLKFSKKDLSCSNFSLLAYQFDHFSHEKIKDLLRKNHSIMQLCNSKNAFVFLQYDKNFIQIRRVFP
Especificaciones

Especificaciones

NP_001695
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
QDFWCSTLVKGVIYGSYSVSEMFPKNFTNCTWTLENPDPTKYSIYLKFSKKDLSCSNFSLLAYQFDHFSHEKIKDLLRKNHSIMQLCNSKNAFVFLQYDKNFIQIRRVFP
RUO
BAI3
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
577
BAI3 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KIAA0550/MGC133100
BAI3
Recombinant
wheat germ expression system
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales