Learn More
Abnova™ Human ATXN10 Partial ORF (NP_037368, 1 a.a. - 109 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00025814-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Description
The autosomal dominant cerebellar ataxias (ADCAs) are a clinically and genetically heterogeneous group of disorders characterized by ataxia, dysarthria, dysmetria, and intention tremor. All ADCAs involve some degree of cerebellar dysfunction and a varying degree of signs from other components of the nervous system. A commonly accepted clinical classification (Harding, 1993) divides ADCAs into 3 different groups based on the presence or absence of associated symptoms such as brainstem signs or retinopathy. The presence of pyramidal and extrapyramidal symptoms and ophthalmoplegia makes the diagnosis of ADCA I, the presence of retinopathy points to ADCA II, and the absence of associated signs to ADCA III. Genetic linkage and molecular analyses revealed that ADCAs are genetically heterogeneous even within the various subtypes.[supplied by OMIM]
Sequence: MAAPRPPPARLSGVMVPAPIQDLEALRALTALFKEQRNRETAPRTIFQRVLDILKKSSHAVELACRDPSQVENLASSLQLITECFRCLRNACIECSVNQNSIRNLDTIGSpecifications
NP_037368 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAAPRPPPARLSGVMVPAPIQDLEALRALTALFKEQRNRETAPRTIFQRVLDILKKSSHAVELACRDPSQVENLASSLQLITECFRCLRNACIECSVNQNSIRNLDTIG | |
RUO | |
ATXN10 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
25814 | |
ATXN10 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
E46L/FLJ37990/SCA10 | |
ATXN10 | |
Recombinant | |
wheat germ expression system |