missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ATP6V1E2 Partial ORF (NP_542384, 129 a.a. - 189 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00090423-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: EPVMIVRCRPQDLLLVEAAVQKAIPEYMTISQKHVEVQIDKEAYLAVNAAGGVEVYSGNQREspecificaciones
NP_542384 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.45kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
EPVMIVRCRPQDLLLVEAAVQKAIPEYMTISQKHVEVQIDKEAYLAVNAAGGVEVYSGNQR | |
RUO | |
ATP6V1E2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
90423 | |
ATP6V1E2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ATP6E1/ATP6EL2/ATP6V1EL2/MGC9341/VMA4 | |
ATP6V1E2 | |
Recombinant | |
wheat germ expression system |