missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ATP6V0D2 Partial ORF (NP_689778, 238 a.a. - 306 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00245972-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: ETLYPTFGKLYPEGLRLLAQAEDFDQMKNVADHYGVYKPLFEAVGGSGGKTLEDVFYEREVQMNVLAFNEspecificaciones
NP_689778 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.33kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
ETLYPTFGKLYPEGLRLLAQAEDFDQMKNVADHYGVYKPLFEAVGGSGGKTLEDVFYEREVQMNVLAFN | |
RUO | |
ATP6V0D2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
245972 | |
ATP6V0D2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ATP6D2/FLJ38708/VMA6 | |
ATP6V0D2 | |
Recombinant | |
wheat germ expression system |