Learn More
Abnova™ Human ATP6AP2 Full-length ORF (NP_005756.2, 1 a.a. - 350 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00010159-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a protein that is associated with adenosine triphosphatases (ATPases). Proton-translocating ATPases have fundamental roles in energy conservation, secondary active transport, acidification of intracellular compartments, and cellular pH homeostasis. There are three classes of ATPases- F, P, and V. The vacuolar (V-type) ATPases have a transmembrane proton-conducting sector and an extramembrane catalytic sector. The encoded protein has been found associated with the transmembrane sector of the V-type ATPases. [provided by RefSeq]
Sequence: MAVFVVLLALVAGVLGNEFSILKSPGSVVFRNGNWPIPGERIPDVAALSMGFSVKEDLSWPGLAVGNLFHRPRATVMVMVKGVNKLALPPGSVISYPLENAVPFSLDSVANSIHSLFSEETPVVLQLAPSEERVYMVGKANSVFEDLSVTLRQLRNRLFQENSVLSSLPLNSLSRNNEVDLLFLSELQVLHDISSLLSRHKHLAKDHSPDLYSLELAGLDEIGKRYGEDSEQFRDASKILVDALQKFADDMYSLYGGNAVVELVTVKSFDTSLIRKTRTILEAKQAKNPASPYNLAYKYNFEYSVVFNMVLWIMIALALAVIITSYNIWNMDPGYDSIIYRMTNQKIRMDEspecificaciones
NP_005756.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
65.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
RUO | |
ATP6AP2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
10159 | |
ATP6AP2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
APT6M8-9/ATP6IP2/ATP6M8-9/ELDF10/HT028/M8-9/MGC99577/MRXE/MSTP009/XMRE | |
ATP6AP2 | |
Recombinant | |
wheat germ expression system |