missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ASB13 Partial ORF (AAH12056, 74 a.a. - 173 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00079754-Q01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16194433

  • 335.00€ / 10 microgramos
Fecha estimada de envío: 27-06-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The protein encoded by this gene is a member of the ankyrin repeat and SOCS box-containing (ASB) family of proteins. They contain ankyrin repeat sequence and a SOCS box domain. The SOCS box serves to couple suppressor of cytokine signalling (SOCS) proteins and their binding partners with the elongin B and C complex, possibly targeting them for degradation. Multiple alternatively spliced transcript variants have been described for this gene, but their full-length sequences are not known. [provided by RefSeq]

Sequence: AGAQVDARNIDGSTPLCDACASGSIECVKLLLSYGAKVNPPLYTASPLHEACMSGSSECVRLLIDVGANLEAHDCHFGTPLHVACAREHLDCVKVLLNAA
Especificaciones

Especificaciones

AAH12056
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
AGAQVDARNIDGSTPLCDACASGSIECVKLLLSYGAKVNPPLYTASPLHEACMSGSSECVRLLIDVGANLEAHDCHFGTPLHVACAREHLDCVKVLLNAA
RUO
ASB13
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
79754
ASB13 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ13134/MGC19879
ASB13
Recombinant
wheat germ expression system
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales