Learn More
Abnova™ Human ARL5B Full-length ORF (NP_848930.1, 1 a.a. - 179 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00221079-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
ARL8 belongs to a family of proteins that are structurally similar to ADP-ribosylation factors (ARFs; see MIM 103180). ARLs and ARFs are part of the RAS superfamily of regulatory GTPases.[supplied by OMIM]
Sequence: MGLIFAKLWSLFCNQEHKVIIVGLDNAGKTTILYQFLMNEVVHTSPTIGSNVEEIVVKNTHFLMWDIGGQESLRSSWNTYYSNTEFIILVVDSIDRERLAITKEELYRMLAHEDLRKAAVLIFANKQDMKGCMTAAEISKYLTLSSIKDHPWHIQSCCALTGEGLCQGLEWMTSRIGVREspecificaciones
NP_848930.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
46.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
RUO | |
ARL5B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
221079 | |
ARL5B (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARL8 | |
ARL5B | |
Recombinant | |
wheat germ expression system |