missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ARHGEF16 Full-length ORF (NP_055263.1, 1 a.a. - 421 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00027237-P02.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Although the specific function of this protein is not known yet, it is thought to be involved in protein-protein and protein-lipid interactions. [provided by RefSeq]
Sequence: MFEILTSEFSYQHSLSILVEEFLQSKELRATVTQMEHHHLFSNILDVLGASQRFFEDLEQRHKAQVLVEDISDILEEHAEKYFHPYIAYCSNEVYQQRTLQKLISSNAAFREALREIERRPACGGLPMLSFLILPMQRVTRLPLLMDTLCLKTQGHSERYKAASRALKAISKLVRQCNEGAHRMERMEQMYTLHTQLDFSKVKSLPLISASRWLLKRGELFLVEETGLFRKIASRPTCYLFLFNDVLVVTKKKSEESYMVQDYAQMNHIQVEKIEPSELPLPGGGNRSSSVPHPFQVTLLRNSEGRQEQLLLSSDSASDRARWIVALTHSERQWQGLSSKGDLPQVEITKAFFAKQADEVTLQQADVVLVLQQEDGWLYGERLRDGETGWFPEDFARFITSRVAVEGNVRRMERLRVETDVEspecificaciones
NP_055263.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
75.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
ARHGEF16 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
27237 | |
ARHGEF16 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GEF16/NBR | |
ARHGEF16 | |
Recombinant | |
wheat germ expression system |