Learn More
Abnova™ Human ARHGEF11 Partial ORF (AAH57394, 651 a.a. - 750 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00009826-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Rho GTPases play a fundamental role in numerous cellular processes that are initiated by extracellular stimuli that work through G protein coupled receptors. The encoded protein may form a complex with G proteins and stimulate Rho-dependent signals. A similar protein in rat interacts with glutamate transporter EAAT4 and modulates its glutamate transport activity. Expression of the rat protein induces the reorganization of the actin cytoskeleton and its overexpression induces the formation of membrane ruffling and filopodia. Two alternative transcripts encoding different isoforms have been described. [provided by RefSeq]
Sequence: RSESLKGREEMKRSRKAENVPRSRSDVDMDAAAEATRLHQSASSSTSSLSTRSLENPTPPFTPKMGRRSIESPSLGFCTDTLLPHLLEDDLGQLSDLEPEEspecificaciones
AAH57394 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RSESLKGREEMKRSRKAENVPRSRSDVDMDAAAEATRLHQSASSSTSSLSTRSLENPTPPFTPKMGRRSIESPSLGFCTDTLLPHLLEDDLGQLSDLEPE | |
RUO | |
ARHGEF11 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9826 | |
ARHGEF11 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp667F1223/GTRAP48/KIAA0380/PDZ-RHOGEF | |
ARHGEF11 | |
Recombinant | |
wheat germ expression system |