missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ANXA6 Full-length ORF (AAH17046, 1 a.a. - 673 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00000309-P01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16191621

  • 508.00€ / 25 microgramos
Fecha estimada de envío: 11-06-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Annexin VI belongs to a family of calcium-dependent membrane and phospholipid binding proteins. Although their functions are still not clearly defined, several members of the annexin family have been implicated in membrane-related events along exocytotic and endocytotic pathways. The annexin VI gene is approximately 60 kbp long and contains 26 exons. It encodes a protein of about 68 kDa that consists of eight 68-amino acid repeats separated by linking sequences of variable lengths. It is highly similar to human annexins I and II sequences, each of which contain four such repeats. Exon 21 of annexin VI is alternatively spliced, giving rise to two isoforms that differ by a 6-amino acid insertion at the start of the seventh repeat. Annexin VI has been implicated in mediating the endosome aggregation and vesicle fusion in secreting epithelia during exocytosis. [provided by RefSeq]

Sequence: MAKPAQGAKYRGSIHDFPGFDPNQDAEALYTAMKGFGSDKEAILDIITSRSNRQRQEVCQSYKSLYGKDLIADLKYELTGKFERLIVGLMRPPAYCDAKEIKDAISGIGTDEKCLIEILASRTNEQMHQLVAAYKDAYERDLEADIIGDTSGHFQKMLVVLLQGTREEDDVVSEDLVQQDVQDLYEAGELKWGTDEAQFIYILGNRSKQHLRLVFDEYLKTTGKPIEASIRGELSGDFEKLMLAVVKCIRSTPEYFAERLFKAMKGLGTRDNTLIRIMVSRSELDMLDIREIFRTKYEKSLYSMIKNDTSGEYKKTLLKLSGGDDDAAGQFFPEAAQVAYQMWELSAVARVELKGTVRPANDFNPDADAKALRKAMKGLGTDEDTIIDIITHRSNVQRQQIRQTFKSHFGRDLMTDLKSEISGDLARLILGLMMPPAHYDAKQLKKAMEGAGTDEKALIEILATRTNAEIRAINEAYKEDYHKSLEDALSSDTSGHFRRILISLATGHREEGGENLDQAREDAQVAAEILEIADTPSGDKTSLETRFMTILCTRSYPHLRRVFQEFIKMTNYDVEHTIKKEMSGDVRDAFVAIVQSVKNKPLFFADKLYKSMKGAGTDEKTLTRIMVSRSEIDLLNIRREFIEKYDKSLHQAIEGDTSGDFLKALLALCGGED
Especificaciones

Especificaciones

AAH17046
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
99.55kDa
Glutathione Sepharose 4 Fast Flow
25 ug
RUO
ANXA6
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
309
ANXA6 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ANX6/CBP68
ANXA6
Recombinant
wheat germ expression system
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales