Learn More
Abnova™ Human ANKRD17 Full-length ORF (AAH43394.1, 1 a.a. - 751 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00026057-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Description
This gene encodes a protein with ankyrin repeats, which are associated with protein-protein interactions. Studies in mice suggest that this protein is involved in liver development. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MEKATVPVAAATAAEGEGSPPAVAAVAGPPAAAEVGGGVGGSSRARSASSPRGMVRVCDLLLKKKPPQQQHHKAKRNRTCRPPSSSESSSDSDNSGGGGGGGGGGGGGGGTSSNNSEEEEDDDDEEEEVSEVESFILDQDDLENPMLETASKLLLSGTADGADLRTVDPETQARLEALLEAAGIGKLSTADGKAFADPEVLRRLTSSVSCALDEAAAALTRMRAESTANAGQSDNRSLAEACSEGDVNAVRKLLIEGRSVNEHTEEGESLLCLACSAGYYELAQVLLAMHANVEDRGIKGDITPLMAAANGGHVKIVKLLLAHKADVNAQSSTGNTALTYACAGGYVDVVKVLLESGASIEDHNENGHTPLMEAGSAGHVEVARLLLENGAGINTHSNEFKESALTLACYKGHLEMVRFLLEAGADQEHKTDEMHTALMEACMDGHVEVARLLLDSGAQVNMPADSFESPLTLAACGGHVELAALLIERGASLEEVNDEGYTPLMEAAREGHEEMVALLLGQGANINAQTEETQETALTLACCGGFLEVADFLIKAGADIELGCSTPLMEAAQEGHLELVKYLLAAGANVHATTATGDTALTYACENGHTDVADVLLQAGADLEHESEGGRTPLMKAARAGHVCTVQFLISKGANVNRTTANNDHTVLSLACAGGHLAVVELLLAHGADPTHRLKDGSTMLIEAAKGGHTSVVCYLLDYPNNLLSAPPPDVTQLTPPSHDLTDNIFPRLVCSpécification
AAH43394.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
104.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
ANKRD17 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
26057 | |
ANKRD17 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ22206/GTAR/KIAA0697/NY-BR-16 | |
ANKRD17 | |
Recombinant | |
wheat germ expression system |