Learn More
Abnova™ Human ALF Full-length ORF (NP_006863.2, 1 a.a. - 478 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00011036-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The assembly and stability of the RNA polymerase II transcription pre-initiation complex on a eukaryotic core promoter involve the effects of TFIIA on the interaction between TATA-binding protein (TBP) and DNA. This gene encodes a germ cell-specific counterpart of the large (alpha/beta) subunit of general transcription factor TFIIA that is able to stabilize the binding of TBP to DNA and may be uniquely important to testis biology. Alternative splicing for this locus has been observed and two variants, encoding distinct isoforms, have been identified. Co-transcription of this gene and the neighboring upstream gene generates a rare transcript (SALF), which encodes a fusion protein comprised of sequence sharing identity with each individual gene product. [provided by RefSeq]
Sequence: MACLNPVPKLYRSVIEDVIEGVRNLFAEEGIEEQVLKDLKQLWETKVLQSKATEDFFRNSIQSPLFTLQLPHSLHQTLQSSTASLVIPAGRTLPSFTTAELGTSNSSANFTFPGYPIHVPAGVTLQTVSGHLYKVNVPIMVTETSGRAGILQHPIQQVFQQLGQPSVIQTSVPQLNPWSLQATTEKSQRIETVLQQPAILPSGPVDRKHLENATSDILVSPGNEHKIVPEALLCHQESSHYISLPGVVFSPQVSQTNSNVESVLSGSASMAQNLHDESLSTSPHGALHQHVTDIQLHILKNRMYGCDSVKQPRNIEEPSNIPVSEKDSNSQVDLSIRVTDDDIGEIIQVDGSGDTSSNEEIGSTRDADENEFLGNIDGGDLKVPEEEADSISNEDSATNSSDNEDPQVNIVEEDPLNSGDDVSEQDVPDLFDTDNVIVCQYDKIHRSKNKWKFYLKDGVMCFGGRDYVFAKAIGDAEWEspecificaciones
NP_006863.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
78.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ALF/MGC26254 | |
GTF2A1L | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
11036 | |
ALF (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MACLNPVPKLYRSVIEDVIEGVRNLFAEEGIEEQVLKDLKQLWETKVLQSKATEDFFRNSIQSPLFTLQLPHSLHQTLQSSTASLVIPAGRTLPSFTTAELGTSNSSANFTFPGYPIHVPAGVTLQTVSGHLYKVNVPIMVTETSGRAGILQHPIQQVFQQLGQPSVIQTSVPQLNPWSLQATTEKSQRIETVLQQPAILPSGPVDRKHLENATSDILVSPGNEHKIVPEALLCHQESSHYISLPGVVFSPQVSQTNSNVESVLSGSASMAQNLHDESLSTSPHGALHQHVTDIQLHILKNRMYGCDSVKQPRNIEEPSNIPVSEKDSNSQVDLSIRVTDDDIGEIIQVDGSGDTSSNEEIGSTRDADENEFLGNIDGGDLKVPEEEADSISNEDSATNSSDNEDPQVNIVEEDPLNSGDDVSEQDVPDLFDTDNVIVCQYDKIHRSKNKWKFYLKDGVMCFGGRDYVFAKAIGDAEW | |
RUO | |
GTF2A1L | |
Wheat Germ (in vitro) | |
GST | |
Liquid |