Learn More
Abnova™ Human ALDH1L2 Full-length ORF (AAI03936.1, 1 a.a. - 488 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00160428-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a protein, which belongs to both the aldehyde dehydrogenase superfamily and the formyl transferase superfamily. Alternatively spliced transcript variants have been found for this gene. [provided by RefSeq]
Sequence: MVTFYGSTLLNSSVPPGEPLEIKGAKKPGLVTKNGLVLFGNDGKALTVRNLQFEDGKMIPASQYFSTGETSVVELTAEEVKVAETIKVIWAGILSNVPIIEDSTDFFKSGASSMDVARLVEEIRQKCGGLQLQNEDVYMATKFEGFIQKVVRKLRGEDQEVELVVDYISKEVNEIMVKMPYQCFINGQFTDADDGKTYDTINPTDGSTICKVSYASLADVDKAVAAAKDAFENGEWGRMNARERGRLMYRLADLLEENQEELATIEALDSGAVYTLALKTHIGMSVQTFRYFAGWCDKIQGSTIPINQARPNRNLTFTKKEPLGVCAIIIPWNYPLMMLAWKSAACLAAGNTLVLKPAQVTPLTALKFAELSVKAGFPKGVINIIPGSGGIAGQRLSEHPDIRKLGFTGSTPIGKQIMKSCAVSNLKKVSLELGGKSPLIIFNDCELDKAVRMGMGAVFFNKGENCIAAGRLFVEESIHDEFVTRVIHEspecificaciones
AAI03936.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
79.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
ALDH1L2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
160428 | |
ALDH1L2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686A16126/DKFZp686M064/FLJ38508/MGC119536/MGC119537 | |
ALDH1L2 | |
Recombinant | |
wheat germ expression system |