Learn More
Abnova™ Human AEBP1 Partial ORF (NP_001120, 912 a.a. - 1013 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00000165-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The adipocyte enhancer binding protein 1 is a transcriptional repressor with carboxypeptidase (CP) activity. This protein binds to a regulatory sequence, adipocyte enhancer 1 (AE-1), located in the proximal promoter region of the adipose P2 (aP2) gene, which encodes the adipocyte fatty-acid binding protein. It is characterized as a member of the regulatory B-like CP family. This protein seems to be activated by a novel mechanism, whereby the direct binding of DNA enhances its protease activity. Adipocyte-enhancer binding protein 1 may play a role in differentiated vascular smooth muscle cells. [provided by RefSeq]
Sequence: VTDEQGIPIANATISVSGINHGVKTASGGDYWRILNPGEYRVTAHAEGYTPSAKTCNVDYDIGATQCNFILARSNWKRIREIMAMNGNRPIPHIDPSRPMTPEspecificaciones
NP_001120 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.96kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VTDEQGIPIANATISVSGINHGVKTASGGDYWRILNPGEYRVTAHAEGYTPSAKTCNVDYDIGATQCNFILARSNWKRIREIMAMNGNRPIPHIDPSRPMTP | |
RUO | |
AEBP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
165 | |
AEBP1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ACLP/FLJ33612 | |
AEBP1 | |
Recombinant | |
wheat germ expression system |